Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR/TNFRSF18 Antibody (T9P4G4*D7), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250337AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250337AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250337AF Supplier Novus Biologicals Supplier No. NBP250337AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen GITR/TNFRSF18
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone T9P4G4*D7
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Gene Symbols TNFRSF18
Host Species Mouse
Immunogen Peptide Sequence - TEDARSCQFP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8784
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.