Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Givinostat hydrochloride

Click to view available options
Quantity:
5 mg
25 mg
Description
Givinostat hydrochloride is a histone deacetylase (HDAC) inhibitor (IC50 = 7.5 to 16 nM). At low concentrations reduces release of pro-inflammatory cytokines, including TNFα, INFγ, IL-1α and IL-1β, from LPS stimulated PBMCs in vitro. Exhibits anti-inflammatory effects in vivo. Improves functional recovery and attenuates tissue damage in a mouse model of traumatic brain injury.
Specifications
Specifications
Chemical Name or Material | [6-[(Diethylamino)methyl]-2-naphthalenyl]methyl-N-[4-[(hydroxyamino)carbonyl]phenyl]carbamate hydrochloride |
CAS | 199657-29-9 |
Assay Percent Range | >98% |
Quantity | 25 mg |
Molecular Formula | C24H27N3O4.HCl |
Formula Weight | Observed MW: 457.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction