Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GKAP/DLGAP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB321735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321735 Supplier Novus Biologicals Supplier No. NBP298138MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GKAP/DLGAP1 Polyclonal antibody specifically detects GKAP/DLGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen GKAP/DLGAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DAP1, DAP-1-ALPHA, DAP-1DAP-1-BETA, discs, large (Drosophila) homolog-associated protein 1, disks large-associated protein 1, GKAPMGC88156, hGKAP, PSD-95/SAP90 binding protein 1, PSD-95/SAP90-binding protein 1, SAP90/PSD-95-associated protein 1, SAPAP1Guanylate kinase-associated protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GKAP/DLGAP1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9229
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.