Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GKN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563479
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA563479 100 μL
1 options

Catalog No. PIPA563479

Supplier: Invitrogen™ PA563479

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPLNNLQWYI YEKQALDNMF SSKYTWVKYN PLESLIKDVD WFLLGSPIEK LCKHIPLYKG EVVENT Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 65%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GKN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GKN2
Gene Accession No. Q86XP6
Gene Alias 1810036H07Rik; Blot; Blottin; BRICD1B; BRICHOS domain containing 1B; Down-regulated in gastric cancer; down-regulated in gastric cancer GDDR; gastrokine 2; gastrokine-2; GDDR; Gkn2; PRO813; RGD1311934; TFIZ1; trefoil factor interactions(z) 1; UNQ465/PRO813; VLTI465
Gene Symbols GKN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GKN2. Recombinant protein control fragment (Product #RP-99699).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.