Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GL7 (T and B Cell Activation Marker) Antibody (GL-7), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200362500UG
Description
GL7 (T and B Cell Activation Marker) Monoclonal specifically detects GL7 (T and B Cell Activation Marker) in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| GL7 (T and B Cell Activation Marker) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgM |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| GL-7 | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| The immunogen for this antibody is T- and B-cell activation antigen (GL7, Ly-77). | |
| 500 μg | |
| Primary | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction