Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Glass Buchner Funnel - Plain Stem
SCI_ED

Catalog No. S41663 Shop All Eisco Products
Click to view available options
Capacity (Metric):
35 mL
80 mL
200 mL
500 mL
1000 mL
Diameter (Metric)Top:
3.2 cm
4.4 cm
7 cm
9 cm
12 cm

Glass Buchner funnels with plain stems and G-3 sintered disks

  • Made from high quality borosilicate glass
  • Includes G-3 sintered disk (15-40 microns)
  • Plain Stem
  • Autoclavable

Warranty and Services

1 year limited manufacturer warranty

Specifications

Diameter (Metric)Top 4.4 cm
Capacity (Metric) 80 mL
Material Borosilicate Glass
Autoclavable Autoclavable
Color Clear
For Use With (Application) filtering liquids
Quantity 1
Weight (Metric) 0.05
Height (English) 7.5
Product Type G-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.