Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Glass Jointed Powder Funnels
SCI_ED

Catalog No. S89492 Shop All Eisco Products
Change view
Click to view available options
Diameter (Metric)Top:
45 mm
55 mm
70 mm
100 mm
Joint Number:
14/23
29/32
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Joint Number
S89492 100 mm 14/23
S89488 45 mm 14/23
S89489 55 mm 14/23
S89490 70 mm 14/23
S89491 70 mm 29/32
S89493 100 mm 29/32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S89492 Supplier Eisco™ Supplier No. CH0460E
Only null left
Add to Cart
Add to Cart

Ideal for transferring powder reagents between pieces of labware. Eisco™ Glass Jointed Powder Funnels feature ground joints to ensure tight connections with other pieces of glassware.

  • Made from high quality borosilicate 3.3 glass
  • Designed with ground joints to ensure tight connections with other pieces of glassware
  • Ideal for transferring powder reagents
  • 60° angle stem
  • Autoclavable

Warranty and Services

1 year limited manufacturer warranty

Specifications

Diameter (Metric)Top 100 mm
Material Borosilicate Glass
Height (Metric) 12.7 cm
Angle 60°
Autoclavable Autoclavable
Color Clear
Description Uniform walls
Graduated No
For Use With (Application) Transferring liquids
Joint Number 14/23
Length (Metric) Stem 55.8 mm
Quantity 1
Length (English) Overall 4 in.
Length (Metric) Overall 10.16 cm
Height (English) 5 in.
Length (English) Stem 2.2 in.
No. of Pores 1
Product Type Filling Funnel
Material (Membrane) Glass
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.