Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLE1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB356897 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356897 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356897 Supplier Novus Biologicals Supplier No. NBP176979MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLE1 Polyclonal antibody specifically detects GLE1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GLE1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias GLE1 (yeast homolog)-like, RNA export mediator, GLE1 RNA export mediator homolog (yeast), GLE1LGLE1 RNA export mediator-like (yeast), GLE1-like protein, GLE1-like, RNA export mediator, hGLE1GLE1 RNA export mediator (yeast), LCCS, LCCS1, lethal congenital contracture syndrome 1, nucleoporin GLE1
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the center of human Gle1. The immunogen is located within amino acids 390 - 440 of Gle1. Amino Acid Squence: NSKDSQAKKIKMDL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2733
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.