Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB600600X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600600X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600600X Supplier Novus Biologicals Supplier No. NB600600AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLI-1 Polyclonal specifically detects GLI-1 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockdown Validated.

Specifications

Antigen GLI-1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot, KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P08151
Gene Alias GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
Gene Symbols GLI1
Host Species Rabbit
Immunogen A synthetic peptide made toward the C-terminus portion of the human GLI-1 protein (between residues 800-850) [Uniprot: P08151]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2735
Test Specificity This is directed against Gli1 from mouse brain. No reaction occurs with human or mouse Gli2 or Gli3.
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.