Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224662 0.1 mg
NBP224662SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224662 Supplier Novus Biologicals Supplier No. NBP224662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLI-1 Polyclonal specifically detects GLI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GLI-1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Alias GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
Gene Symbols GLI1
Host Species Rabbit
Immunogen A portion of amino acids 940-990 of human GLI1 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2735
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human and rhesus monkey, 94% homologous in horse, 88% homologous in dog, and 76% homologous in cow, mouse and rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.