Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-1 Antibody (OTI4E2), FITC, Novus Biologicals™
SDP

Catalog No. NBP270831F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270831F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270831F Supplier Novus Biologicals Supplier No. NBP270831F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLI-1 Monoclonal specifically detects GLI-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Knockdown Validated.

Specifications

Antigen GLI-1
Applications Immunohistochemistry, KnockDown
Classification Monoclonal
Clone OTI4E2
Conjugate FITC
Dilution Immunohistochemistry, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
Gene Symbols GLI1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 755-1106 of human GLI-1 (NP_005260) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2735.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.