Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-2 Antibody (524409), Alexa Fluor 350, Novus Biologicals™
SDP

Catalog No. NB003120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003120 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003120 Supplier Novus Biologicals Supplier No. MAB35262AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLI-2 Monoclonal specifically detects GLI-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen GLI-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 524409
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2
Gene Symbols GLI2
Host Species Mouse
Immunogen E. coli-derived partial recombinant human GLI-2 protein (amino acids 599-948)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cell Cycle and Replication, Neuroscience, Stem Cell Signaling Pathway
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.