Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB151550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151550 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151550 Supplier Novus Biologicals Supplier No. NB600874AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLI-2 Polyclonal antibody specifically detects GLI-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow

Specifications

Antigen GLI-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2
Host Species Rabbit
Immunogen This affinity purified GLI-2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 30-65 of human GLI-2 (isoform a). (Uniprot: P10070)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cell Cycle and Replication, Neuroscience, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2736
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.