Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB005457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005457 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005457 Supplier Novus Biologicals Supplier No. NBP229627AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLI-3 Polyclonal specifically detects GLI-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen GLI-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3
Gene Symbols GLI3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Gli3 protein (between residues 50-120) [UniProt P10071]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Signaling Pathway
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.