Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI4 Antibody (OTI3A9), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289333 Supplier Novus Biologicals Supplier No. NBP270833APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLI4 Monoclonal antibody specifically detects GLI4 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen GLI4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3A9
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias GLI family zinc finger 4, GLI-Kruppel family member GLI4, glioma-associated oncogene family zinc finger 4, HKR4GLI-Kruppel family member GLI4 (oncogene HKR4), Krueppel-related zinc finger protein 4, Protein HKR4, zinc finger protein GLI4, ZNF928
Host Species Mouse
Immunogen Full length human recombinant protein of human GLI4 (NP_612474) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2738
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.