Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI4 Antibody (OTI3A9), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP270833C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270833C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270833C Supplier Novus Biologicals Supplier No. NBP270833C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLI4 Monoclonal specifically detects GLI4 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen GLI4
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3A9
Conjugate DyLight 650
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GLI family zinc finger 4, GLI-Kruppel family member GLI4, glioma-associated oncogene family zinc finger 4, HKR4GLI-Kruppel family member GLI4 (oncogene HKR4), Krueppel-related zinc finger protein 4, Protein HKR4, zinc finger protein GLI4, ZNF928
Gene Symbols GLI4
Host Species Mouse
Immunogen Full length recombinant protein of human GLI4 (NP_612474) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2738.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.