Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLIPR2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP247126C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247126C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247126C Supplier Novus Biologicals Supplier No. NBP247126C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLIPR2 Polyclonal specifically detects GLIPR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GLIPR2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C9orf19Glioma pathogenesis-related protein 2, chromosome 9 open reading frame 19, GAPR1, GAPR-117kD fetal brain protein, GLI pathogenesis-related 2, GliPR 2, Golgi-associated plant pathogenesis-related protein 1, Golgi-associated PR-1 protein
Gene Symbols GLIPR2
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human GAPR1 protein (between amino acids 90-154) [UniProt Q9H4G4]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 152007
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.