Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GliS2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396407 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396407 0.025 mg
NBP241311 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396407 Supplier Novus Biologicals Supplier No. NBP2413110.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GliS2 Polyclonal specifically detects GliS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GliS2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias GLIS family zinc finger 2, NKL, zinc finger protein GLIS2
Gene Symbols GLIS2
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the center of human GLIS2. The immunogen is located within amino acids 260 - 310 of GLIS2. Amino Acid Squence: NKRYSNSSDRFKHTRTH
Molecular Weight of Antigen 60 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84662
Test Specificity GLIS2 antibody is human, mouse and rat reactive.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.