Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLIS3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216668 Supplier Novus Biologicals Supplier No. NBP216668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLIS3 Polyclonal specifically detects GLIS3 in Human samples. It is validated for Western Blot.

Specifications

Antigen GLIS3
Applications Western Blot
Classification Polyclonal
Concentration 0.87mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8NEA6
Gene Alias FLJ38999, FLJ90578, GLIS family zinc finger 3, GLI-similar 3, OK/KNS-cl.4, zinc finger protein 515, zinc finger protein GLIS3
Gene Symbols GLIS3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human GLIS3. The exact sequence is proprietary.
Molecular Weight of Antigen 84 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169792
Test Specificity Expected to cross react based on sequence identity: Mouse (88%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.