Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Globe Scientific Universal Snap Caps
Click to view available options
Color:
Black
Blue
Gray
Green
Lavender
Natural
Red
Yellow
Description
- Precision molded low-density polyethylene made
- Fits most 12, 13 and 16mm plastic test tubes
- Avoids cross-contamination, protects lab work areas against infection and spillage
- Not autoclavable
- Assorted colors available for sample identification
Specifications
Specifications
| Product Type | Snap Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Green |
| Size | Universal |
| For Use With (Equipment) | Recapping 12, 13 and 16 mm evacuated glass blood collection test tubes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction