Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globo H Antibody (VK9), Novus Biologicals™
SDP

Catalog No. NB036545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036545 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036545 Supplier Novus Biologicals Supplier No. NBP288954
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Globo H Monoclonal specifically detects Globo H in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Globo H
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone VK9
Conjugate Unconjugated
Formulation 0.2 uM filtered solution PBS, pH 7.2
Gene Alias DR3, Fuca1-2Galb1-3GlcNAcb, Fucalpha1-2Galbeta1-3GlcNAcbeta, globo-H, H type I epitope, H TYPE I TRISACCHARIDE, human blood group H type 1 trisaccharide
Host Species Mouse
Immunogen Globo H-KLH (keyhole limpet hemocyanin) synthetic conjugate
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.