Learn More
Description
Includes
NL493-conjugated Mouse Anti-Human SSEA-4, NL557-conjugated Mouse Anti-Human TRA-1-60™, NL493-conjugated Mouse Anti-Human TRA-1-81, NL557-conjugated Mouse Anti-Human SSEA-1, 50X stock,
Embryonic stem (ES) cells are pluripotent stem cells derived from the inner cell mass of pre-implantation embryos. Induced pluripotent stem (iPS) cells can be generated by somatic cell reprogramming following the exogenous expression of specific transcription factors (Oct-3/4, KLF4, SOX2, and c-Myc). These cell types are capable of unlimited, undifferentiated proliferation in vitro and still maintain the capacity to differentiate into a wide variety of somatic cells. In this capacity, pluripotent stem cells have widespread clinical potential for the treatments of heart disease, diabetes, spinal cord injury, and a variety of neurodegenerative disorders.
Benefits:
- Verify human stem cell pluripotency in 30 minutes
- Includes 4 pluripotent stem cell-specific fluorochrome-conjugated antibodies
- Single-step protocol
- No adverse effects on stemness or proliferation
- Pick pluripotent human stem cell colonies with increased confidence
Storage: Store unopened product at 2° to 8°C. Do not use past expiration date.
Alternate Names: Pluripotent Stem Cells
Specifications
Specifications
| Cell Type | Pluripotent Stem cells |
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Description | Derived from Embryonic stem (ES) Cells |
| For Use With (Application) | Cell Imaging, Verification Stem Cell Pluripotency |
| Product Type | Live Cell Imaging Kit |
| Quantity | 1 Kit |
| Species | Human |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
