Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP-1R Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NLS1205C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NLS1205C 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NLS1205C Supplier Novus Biologicals Supplier No. NLS1205C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLP-1R Polyclonal specifically detects GLP-1R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GLP-1R
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot 1.0 μg/mL, Immunohistochemistry 10-20 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-20 μg/mL
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P43220
Gene Alias GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331
Gene Symbols GLP1R
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal extracellular portion of the human GLP1R protein (between residues 100-200). [UniProt P43220]
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2740
Test Specificity Human and mouse. The immunogen used for this antibody has 100/88% homology/identity with the rat protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.