Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP-1R Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB337742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337742 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337742 Supplier Novus Biologicals Supplier No. NBP197308PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLP-1R Polyclonal antibody specifically detects GLP-1R in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, KnockDown

Specifications

Antigen GLP-1R
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated
Formulation PBS
Gene Alias GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2740
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.