Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP-2R Antibody (413801), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB370388 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370388 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370388 Supplier Novus Biologicals Supplier No. FAB4285Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLP-2R Monoclonal specifically detects GLP-2R in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen GLP-2R
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 413801
Conjugate DyLight 755
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias GLP-2 receptor, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human GLP-2R, Met1-Ile553, Accession # O95838
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9340
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.