Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP-2R Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen GLP-2R
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159029
SDP
View Documents
Novus Biologicals
NBP159029
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

GLP-2R Polyclonal specifically detects GLP-2R in Human samples. It is validated for Western Blot.
Specifications

Specifications

GLP-2R
Polyclonal
Rabbit
Cell Cycle and Replication, GPCR
O95838
9340
Synthetic peptides corresponding to GLP2R(glucagon-like peptide 2 receptor) The peptide sequence was selected from the N terminal of GLP2R. Peptide sequence KLGSSRAGPGRGSAGLLPGVHELPMGIPAPWGTSPLSFHRKCSLWAPGRP.
Primary
Western Blot
Unconjugated
RUO
Human
GLP-2 receptor, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor
GLP2R
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.