Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP/EHMT1 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP177400V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177400V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177400V Supplier Novus Biologicals Supplier No. NBP177400V

Rabbit Polyclonal Antibody

GLP/EHMT1 Polyclonal specifically detects GLP/EHMT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen GLP/EHMT1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9H9B1
Gene Alias EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 1H3-K9-HMTase 5, Eu-HMTase1DEL9q34, EUHMTASE1FP13812, FLJ12879, G9a-like protein 1, GLP, GLP1, H3 lysine-9 specific 5, KIAA1876euchromatic histone methyltransferase 1, KMT1DDKFZp667M072
Gene Symbols EHMT1
Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human EHMT1 protein (within residues 20-200). [Swiss-Prot Q9H9B1]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 79813
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.