Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP1 Antibody (03), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP221747AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221747AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221747AH Supplier Novus Biologicals Supplier No. NBP221747AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLP1 Monoclonal specifically detects GLP1 in Mammal samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GLP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1(9-37) adsorbed onto aluminum hydroxide gel.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity binds GLP-1(1-37), GLP-1(7-37) and GLP-1(9-37). Cross-reaction with C-terminally amidated GLP-1 is 0-16% in various circumstances.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.