Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP1 Antibody (10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP105180AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105180AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105180AF Supplier Novus Biologicals Supplier No. NBP105180AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLP1 Monoclonal specifically detects GLP1 in Mammal samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GLP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity This antibody binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(9-36)amide and human GIP. It ross-reacts 0.25% with GLP-1(1-37), approximately 6% with human GLP-2 and approximately 50% with exendin-4.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.