Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP1 Antibody (4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2238A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2238A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2238A30 Supplier Novus Biologicals Supplier No. NBP223558AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLP1 Monoclonal specifically detects GLP1 in Human, Mouse, Mammal samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen GLP1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4
Conjugate Alexa Fluor 350
Dilution ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity This antibody binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(1-37), GLP-1(9-36)amide, human GIP and exendin-4. Cross-reacts approximately 1% with human GLP-2.
Target Species Human, Mouse, Mammalia
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.