Learn More
Description
Specifications
Specifications
| Antigen | GLP1 |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Formulation | 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide |
| Gene Alias | glicentin-related polypeptide, GLP1, GLP2, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP |
| Gene Symbols | GCG |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
