Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Antibody (4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$207.00 - $486.50
Specifications
Antigen | GLP1 |
---|---|
Clone | 4 |
Concentration | 1 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
Applications | Immunocytochemistry, Immunofluorescence, ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019331
![]() |
Novus Biologicals
NBP2235580.025ML |
0.025 mL |
Each for $207.00
|
|
|||||
NBP22355801
![]() |
Novus Biologicals
NBP2235580.1ML |
0.1 mL |
Each for $486.50
|
|
|||||
Description
GLP1 Monoclonal specifically detects GLP1 in Human, Mouse, Mammal samples. It is validated for ELISA, Immunohistochemistry.Specifications
GLP1 | |
1 mg/mL | |
Immunocytochemistry, Immunofluorescence, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Mammalia, Mouse | |
glicentin-related polypeptide, GLP1, GLP2, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
GCG | |
IgG1 κ | |
Protein A or G purified | |
Clone 4 binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(1-37), GLP-1(9-36)amide, glucagon, human GIP and exendin-4. Cross-reacts approximately 1% with human GLP-2. |
4 | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
Monoclonal | |
Purified | |
Cancer, GPCR, Signal Transduction | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
2641 | |
Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title