Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP1 Mouse anti-Human, Mammal, Clone: 1100, Novus Biologicals™
SDP

Catalog No. NB019332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019332 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019332 Supplier Novus Biologicals Supplier No. NBP2235610.025ML

Mouse Monoclonal Antibody

GLP1 Monoclonal specifically detects GLP1 in Human, Mammal samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GLP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1100
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity Reacts with all forms of GLP-1, including precursor and GLP-1(9-37)GLP-1(9-36amide) metabolite
Target Species Mammalia, Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.