Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Mouse anti-Human, Mammal, Clone: 1100, Novus Biologicals™

Mouse Monoclonal Antibody
$206.00
Specifications
| Antigen | GLP1 |
|---|---|
| Clone | 1100 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
Description
GLP1 Monoclonal specifically detects GLP1 in Human, Mammal samples. It is validated for Western Blot, ELISA.Specifications
| GLP1 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Mammalia, Human | |
| GCG | |
| IgG2a κ | |
| Protein A or G purified |
| 1100 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Cancer, GPCR, Signal Transduction | |
| 2641 | |
| Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title