Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLS2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB184642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184642 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184642 Supplier Novus Biologicals Supplier No. NBP282071JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLS2 Polyclonal antibody specifically detects GLS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GLS2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.2, GAbreast cell glutaminase, GLSglutaminase GA, glutaminase 2 (liver, mitochondrial), glutaminase I, glutaminase liver isoform, mitochondrial, hLGA, LGA, L-glutaminase, L-glutamine amidohydrolase, MGC71567, phosphate-activated glutaminase, phosphate-dependent glutaminase, truncated glutaminase 2
Host Species Rabbit
Immunogen GLS2 antibody was raised against an 18 amino acid synthetic peptide near the center terminus of human GLS2. The immunogen is located within amino acids 300 - 350 of GLS2. Amino Acid Squence: NATFQSEKETGDRNYAIG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27165
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.