Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glu-Glu Epitope Tag Antibody, Agarose, Novus Biologicals™
SDP

Catalog No. NB600360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600360 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600360 Supplier Novus Biologicals Supplier No. NB600360

Goat Polyclonal Antibody

Glu-Glu Epitope Tag Polyclonal antibody specifically detects Glu-Glu Epitope Tag in Epitope Tag samples. It is validated for Immunoprecipitation.

Specifications

Antigen Glu-Glu Epitope Tag
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Agarose
Dilution Immunoprecipitation 1:10-1:500
Formulation PBS with 0.09% Sodium Azide
Gene Alias EYMPME epitope tag, EYMPME tag, Glu-Glu epitope tag
Host Species Goat
Immunogen Goats were immunized with a synthetic peptide representing amino acid residues 314 - 319 (EYMPME) of middle T antigen of mouse polymavirus conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epitope Tags, glu glu Epitope Tags
Primary or Secondary Primary
Test Specificity Antibody recognizes the glu-glu cleavage site.
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.