Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glu-Glu Epitope Tag Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600354 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600354 Supplier Novus Biologicals Supplier No. NB600354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glu-Glu Epitope Tag Polyclonal antibody specifically detects Glu-Glu Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Glu-Glu Epitope Tag
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:30000, ELISA Primary-1:1000-1:30000; Coating: 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunoprecipitation 1 - 4 ug/mg lysate
Formulation PBS with 0.09% Sodium Azide
Gene Alias EYMPME epitope tag, EYMPME tag, Glu-Glu epitope tag
Host Species Rabbit
Immunogen Rabbits were immunized with glu-glu cleavage site (EYMPME) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. Rabbit anti- human glu-glu affinity purified antibodies were coupled to peptide immobilized on solid support.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, glu glu Epitope Tags
Primary or Secondary Primary
Test Specificity Antibody recognizes the glu-glu cleavage site.
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.