Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Antibody (08), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP221802A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP221802A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP221802A30 Supplier Novus Biologicals Supplier No. NBP221802AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucagon Monoclonal specifically detects Glucagon in Human, Rat, Porcine samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Antigen Glucagon
Applications ELISA
Classification Monoclonal
Clone 8
Conjugate Alexa Fluor 350
Dilution ELISA, Sandwich ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic N-terminal fragment of glucagon coupled to carrier
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity Clone 08 binds human glucagon
Target Species Human, Rat, Pig
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.