Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Antibody (49), PerCP, Novus Biologicals™
SDP

Catalog No. NB355610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355610 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355610 Supplier Novus Biologicals Supplier No. NBP223591PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucagon Monoclonal antibody specifically detects Glucagon in Mammalia samples. It is validated for Western Blot, ELISA

Specifications

Antigen Glucagon
Applications Western Blot, ELISA
Classification Monoclonal
Clone 49
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Host Species Mouse
Immunogen Synthetic GLP-1(9-18)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Mammalia
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.