Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Antibody (53), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP105184IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105184IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105184IR Supplier Novus Biologicals Supplier No. NBP105184IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucagon Monoclonal specifically detects Glucagon in Mammal samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Glucagon
Applications Western Blot, ELISA
Classification Monoclonal
Clone 53
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1(9-18) adsorbed onto aluminum hydroxide gel
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity Based on inhibition ELISA, It cross-reacts <2% with GLP-1(7-36)amide
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.