Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Mouse anti-Human, Rat, Porcine, Clone: 9, Novus Biologicals™
SDP

Catalog No. NB019260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019260 0.025 mL
NBP221803 0.1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019260 Supplier Novus Biologicals Supplier No. NBP2218030.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Glucagon Monoclonal specifically detects Glucagon in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA, Sandwich ELISA Capture.

Specifications

Antigen Glucagon
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:8000, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin 1:8000, Sandwich ELISA 1:100-1:2000, Sandwich ELISA Capture
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl
Gene Alias glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Host Species Mouse
Immunogen Synthetic C-terminal fragment of glucagon coupled to carrier.
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human, Rat, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.