Learn More
Glucagon Mouse anti-Human, Unconjugated, Clone: 2A8G1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Glucagon |
---|---|
Clone | 2A8G1 |
Concentration | 1000 μg/mL |
Applications | Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9413
![]() ![]() |
Proteintech Group Inc
672861IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The Glucagon antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glucagon. This antibody recognizes human, mouse antigen. The Glucagon antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Glucagon | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
2641 | |
Primary |
2A8G1 | |
Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.