Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon R/GCGR Antibody (591929) [FITC], Novus Biologicals™
SDP

Catalog No. NB401969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401969 Supplier Novus Biologicals Supplier No. FAB10296F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Glucagon R/GCGR
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 591929
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Glucagon R/GCGR, Met1-Gln142, Accession # P47871
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2642
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.