Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon R/GCGR Antibody (9G1), Novus Biologicals™
SDP

Catalog No. NB297862 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297862 50 μL
NB297863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297862 Supplier Novus Biologicals Supplier No. NBP32655750UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for ELISA

Specifications

Antigen Glucagon R/GCGR
Applications ELISA
Classification Monoclonal
Clone 9G1
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
Host Species Human
Immunogen Recombinant Human Glucagon R/GCGR protein [UniProt P47871]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2642
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.