Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon R/GCGR Antibody (hGR-2 F6), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342911 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342911 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342911 Supplier Novus Biologicals Supplier No. NBP281065MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Neutralization

Specifications

Antigen Glucagon R/GCGR
Applications Immunocytochemistry, Neutralization
Classification Monoclonal
Clone hGR-2 F6
Conjugate mFluor Violet 450 SE
Dilution Immunocytochemistry/ Immunofluorescence, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
Host Species Mouse
Immunogen Recombinant human Glucagon R/GCGR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2642
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.