Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucocorticoid Receptor alpha Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300633 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300633 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300633 Supplier Novus Biologicals Supplier No. NB300633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Glucocorticoid Receptor alpha Polyclonal specifically detects Glucocorticoid Receptor alpha in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Glucocorticoid Receptor alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 4 ug/ml, Simple Western, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P04150
Gene Alias GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Gene Symbols NR3C1
Host Species Rabbit
Immunogen Synthetic Peptide: C(755) E I I T N Q I P K Y S N G N I K K(771)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2908
Test Specificity Detects GR alpha from human, mouse and rat tissues. This does not detect GR beta.
Reconstitution Reconstitute with 50 ul ultra-pure water to desired concetration.
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.