Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucono delta-Lactone, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602926
Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

GL107, 90-80-2, C6H10O6

Spectrum™ Chemical Glucono delta-Lactone, FCC is used as a sequestrant, a leavening agent, and an acidifier. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 5 kg
Percent Purity 99 to 100.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.