Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosamine (N-acetyl)-6-Sulfatase/GNS Antibody (007), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB307668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307668 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307668 Supplier Novus Biologicals Supplier No. NBP289413JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glucosamine (N-acetyl)-6-Sulfatase/GNS Monoclonal antibody specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Glucosamine (N-acetyl)-6-Sulfatase/GNS
Applications Western Blot, ELISA
Classification Monoclonal
Clone 007
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.6, EC 3.1.6.14, G6Sglucosamine-6-sulfatase, glucosamine (N-acetyl)-6-sulfatase, Glucosamine-6-sulfatase, MGC21274, N-acetylglucosamine-6-sulfatase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Glucosamine (N-acetyl)-6-Sulfatase/GNS (Accession#: NP_002067.1; Met 1-Leu 552).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2799
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.