Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 1-dehydrogenase Antibody (OTI2A7), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271478R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271478R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271478R Supplier Novus Biologicals Supplier No. NBP271478R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucose 1-dehydrogenase Monoclonal specifically detects Glucose 1-dehydrogenase in Human, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Glucose 1-dehydrogenase
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2A7
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 6-phosphogluconolactonase, DKFZp686A01246, EC 1.1.1.49, EC 2.7.4.3, EC 3.1.1.31, G6PD, H form, G6PDH, GDH/6PGL endoplasmic bifunctional protein, GDHglucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary, hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase), MGC87643
Gene Symbols H6PD
Host Species Mouse
Immunogen Full length recombinant protein of human Glucose 1-dehydrogenase (NP_004276) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9563.0
Target Species Human, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.