Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody (2F6), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP13038A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP13038A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP13038A94 Supplier Novus Biologicals Supplier No. NBP130358AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucose 6 Phosphate Dehydrogenase Monoclonal specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Glucose 6 Phosphate Dehydrogenase
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2F6
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Gene Symbols G6PD
Host Species Mouse
Immunogen Recombinant human G6PD (35-506aa) purified from E. coli
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2539
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.